2,6-Dihydroxy-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
Supplier: TCI America
A1894-1G
A1894-5G
A1894-25G
TCA1894-1GEA
120.71
CAD
TCA1894-1G
TCA1894-5G
TCA1894-25G
2,6-Dihydroxy-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
2,6-Dihydroxy-9,10-anthraquinone
CAS Number: 84-60-6
Molecular Formula: C14H8O4
Molecular Weight: 240.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 330
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2,6-Dihydroxyanthraquinone
Formula:
C₁₄H₈O₄ Melting Pt: 330 °C |
MDL Number:
MFCD00001228 CAS Number: 84-60-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...