1-(1-Naphthyl)ethanol ≥98.0%
Supplier: TCI America
Synonyms:
alpha-Methyl-1-naphthalenemethanol, 1-(1-Hydroxyethyl)naphthalene, (±)-1-(1-Naphthyl)ethanol, 1-(Naphthalen-1-yl)ethanol
N0395-1G
N0395-5G
TCN0395-1GEA
180.12
CAD
TCN0395-1G
TCN0395-005G
1-(1-Naphthyl)ethanol ≥98.0%
1-(1-Naphthyl)ethanol
CAS Number: 1517-72-2
MDL Number: MFCD00016806
Molecular Formula: C12H12O
Molecular Weight: 172.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 64
Other Product Names / Synonyms: 1-(1-Hydroxyethyl)naphthalene, alpha-Methyl-1-naphthalenemethanol
Formula:
C₁₂H₁₂O MW: 172.23 g/mol Boiling Pt: 133 °C Melting Pt: 64 °C |
MDL Number:
MFCD00016806 CAS Number: 1517-72-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...