N-Nitrosodibutylamine ≥97.0%

Supplier: TCI America

Synonyms: N-Dibutylnitrosamine

N0375-25ML N0375-5ML
TCN0375-025MLEA 376.83 CAD
TCN0375-025ML TCN0375-5ML
N-Nitrosodibutylamine ≥97.0%
N-Nitrosodibutylamine

CAS Number: 924-16-3
MDL Number: MFCD00013892
Molecular Formula: C8H18N2O
Molecular Weight: 158.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Specific Gravity (20/20): 0.90


Product Categories: Bioscience, Reagents for Carcinogenic Models
Other Product Names / Synonyms: N-Dibutylnitrosamine

Formula: C₈H₁₈N₂O
Boiling Pt: 237 °C
Density: 0.90 (20 °C)
MDL Number: MFCD00013892
CAS Number: 924-16-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=102c29aea5325766f4eb03fa65ced4b955120d5d&vid=102c29aea5325766f4eb03fa65ced4b955120d5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR