Sodium-2-nitroaniline-4-sulfonate ≥98.0% (by HPLC)

Supplier: TCI America

N0371-500G N0371-25G N0371-100G
TCN0371-500GEA 567.22 CAD
TCN0371-500G TCN0371-025G TCN0371-100G
Sodium-2-nitroaniline-4-sulfonate ≥98.0% (by HPLC)
Sodium-2-nitroaniline-4-sulfonate

CAS Number: 5042-33-1
MDL Number: MFCD00070599
Molecular Formula: C6H6N2O5S
Molecular Weight: 240.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 4-Amino-3-nitrobenzenesulfonic Acid Sodium Salt, 2-Nitroaniline-4-sulfonic Acid Sodium Salt, Sodium 4-Amino-3-nitrobenzenesulfonate

Formula: C₆H₆N₂O₅S
MDL Number: MFCD00070599
CAS Number: 5042-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c4746c3bda8c5d97fc60e8dcbbb63e444138deb&vid=6c4746c3bda8c5d97fc60e8dcbbb63e444138deb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR