4-Bromo-1-naphthylamine ≥98.0%

Supplier: TCI America

A1886-25G A1886-5G
TCA1886-25GEA 262.75 CAD
TCA1886-25G TCA1886-5G
4-Bromo-1-naphthylamine ≥98.0%
4-Bromo-1-naphthylamine

CAS Number: 2298-07-9
Molecular Formula: C10H8BrN
Molecular Weight: 222.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: 4-Bromo-1-naphthylamine

Formula: C₁₀H₈BrN
Melting Pt: 102 °C
MDL Number: MFCD00004023
CAS Number: 2298-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4eb815e8ed6153b5b96ff89e86954fb88fce4718&vid=4eb815e8ed6153b5b96ff89e86954fb88fce4718&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR