1-Nitro-2-naphthol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0370-5G N0370-1G
TCN0370-005GEA 2491.66 CAD
TCN0370-005G TCN0370-001G
1-Nitro-2-naphthol ≥98.0% (by GC, titration analysis)
1-Nitro-2-naphthol

CAS Number: 550-60-7
MDL Number: MFCD00021461
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 103


Other Product Names / Synonyms: 2-Hydroxy-1-nitronaphthalene

Formula: C₁₀H₇NO₃
Melting Pt: 103 °C
MDL Number: MFCD00021461
CAS Number: 550-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d0848a6536b57366b5db38e53ef8ac416ad4e3e&vid=0d0848a6536b57366b5db38e53ef8ac416ad4e3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR