Hexyl nicotinate ≥98.0%

Supplier: TCI America

Synonyms: Nicotinic Acid Hexyl Ester

N0329-25G N0329-5G
TCN0329-025GEA 203.35 CAD
TCN0329-025G TCN0329-005G
Hexyl nicotinate ≥98.0%
Hexyl nicotinate

CAS Number: 23597-82-2
MDL Number: MFCD00023587
Molecular Formula: C12H17NO2
Molecular Weight: 207.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Nicotinic Acid Hexyl Ester

Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Boiling Pt: 140 °C
Density: 1.02 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00023587
CAS Number: 23597-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=219cea2b86152f836b1a6a5cd16596b0b349cd6b&vid=219cea2b86152f836b1a6a5cd16596b0b349cd6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR