4-Acetamidopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-(Pyridin-4-yl)acetamide, N-(4-Pyridyl)acetamide

A1883-25G A1883-5G
TCA1883-25GEA 335.59 CAD
TCA1883-25G TCA1883-5G
4-Acetamidopyridine ≥98.0% (by GC, titration analysis)
4-Acetamidopyridine

CAS Number: 5221-42-1
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 152


Other Product Names / Synonyms: N-(4-Pyridyl)acetamide

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00720126
CAS Number: 5221-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44dc55fc3ceb6a334fb02ec02e6f585561ab816c&vid=44dc55fc3ceb6a334fb02ec02e6f585561ab816c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR