Basic Blue 24 (New Methylene Blue)

Supplier: TCI America

Synonyms: New Methylene Blue N, zinc chloride double salt, New Methylene Blue N

N0318-5G N0318-25G
TCN0318-005GEA 112.18 CAD
TCN0318-005G TCN0318-025G
Basic Blue 24 (New Methylene Blue)
Basic Blue 24 (New Methylene Blue)

CAS Number: 6586-05-6
MDL Number: MFCD00135878
Molecular Formula: C18H22ClN3S
Form: Crystal
Color: Deep Yellow


Other Product Names / Synonyms: New Methylene Blue

Formula: C₁₈H₂₂ClN₃S
MW: 416.05 g/mol
MDL Number: MFCD00135878
CAS Number: 6586-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=953b240d4cb2f67b6dc52eca5c6d9b40982981ca&vid=953b240d4cb2f67b6dc52eca5c6d9b40982981ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR