Nile blue chloride (Basic blue 12)
Supplier: TCI America
N0317-25G
N0317-5G
TCN0317-025GEA
311.26
CAD
TCN0317-025G
TCN0317-5G
Nile blue chloride (Basic blue 12)
Nile blue chloride (Basic blue 12)
CAS Number: 2381-85-3
MDL Number: MFCD00011936
Molecular Formula: C20H20ClN3O
Molecular Weight: 353.85
Form: Crystal
Color: Deep Yellow Green
Other Product Names / Synonyms: Nile Blue A
Formula:
C₂₀H₂₀ClN₃O |
MDL Number:
MFCD00011936 CAS Number: 2381-85-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...