tert-Nonyl Mercaptan (mixture) ≥97.0% (by titrimetric analysis)

Supplier: TCI America

N0299-500ML N0299-25ML
TCN0299-500MLEA 180.12 CAD
TCN0299-500ML TCN0299-025ML
tert-Nonyl Mercaptan (mixture) ≥97.0% (by titrimetric analysis)
tert-Nonyl mercaptan(mixture)

CAS Number: 25360-10-5
MDL Number: MFCD00059963
Molecular Formula: C9H20S
Molecular Weight: 160.32
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Flash Point (°C): 64
Specific Gravity (20/20): 0.85

Formula: C₉H₂₀S
Density: 0,85 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00059963
CAS Number: 25360-10-5

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR