(±)-γ-Nonalactone ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
gamma-Nonalactone, (±)-Nonan-4-olide, gamma-Nonanolactone, 5-Pentyldihydrofuran-2(3H)-one, gamma-Pelargonolactone
CAS Number: 104-61-0
MDL Number: MFCD00005403
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 243
Flash Point (°C): 132
Specific Gravity (20/20): 0.97
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: gamma-Nonalactone, gamma-Pelargonolactone
Formula:
C₉H₁₆O₂ Boiling Pt: 243 °C Density: 0.97 (20 °C) Flash Pt: 132 °C |
MDL Number:
MFCD00005403 CAS Number: 104-61-0 EINECS: 203-219-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...