4-Nitropyrocatechol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

N0273-5G N0273-25G
TCN0273-005GEA 72.35 CAD
TCN0273-005G TCN0273-025G
4-Nitropyrocatechol ≥98.0% (by GC, titration analysis)
4-Nitropyrocatechol

CAS Number: 3316-09-4
MDL Number: MFCD00007242
Molecular Formula: C6H5NO4
Molecular Weight: 155.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 175


Other Product Names / Synonyms: 4-Nitropyrocatechol

Formula: C₆H₅NO₄
Melting Pt: 175 °C
MDL Number: MFCD00007242
CAS Number: 3316-09-4
EINECS: 222-009-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b388c0f0664daa5d53e12e451ed61162bc19e5de&vid=b388c0f0664daa5d53e12e451ed61162bc19e5de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR