4-Hydroxy-3-nitroso-1-naphthalenesulfonic acid hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Nitroso-1-naphthol-4-sulfonic acid, 2-Nitroso-1-naphthol-4-sulphonic acid

N0269-25G N0269-5G
TCN0269-025GEA 246.48 CAD
TCN0269-025G TCN0269-5G
4-Hydroxy-3-nitroso-1-naphthalenesulfonic acid hydrate ≥98.0% (by titrimetric analysis)
4-Hydroxy-3-nitroso-1-naphthalenesulfonic acid hydrate

CAS Number: 3682-32-4
MDL Number: MFCD00003933
Molecular Formula: C10H7NO5S
Molecular Weight: 253.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 145


Other Product Names / Synonyms: 2-Nitroso-1-naphthol-4-sulfonic Acid

Formula: C₁₀H₇NO₅S
MW: 253.23 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00003933
CAS Number: 3682-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd4288d64c9aa78b8f5577cd661c2e39676f98ae&vid=bd4288d64c9aa78b8f5577cd661c2e39676f98ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR