4-Nitro-1-naphthylamine ≥98.0%
Supplier: TCI America
N0213-5G
N0213-25G
TCN0213-005GEA
232.26
CAD
TCN0213-005G
TCN0213-025G
4-Nitro-1-naphthylamine ≥98.0%
4-Nitro-1-naphthylamine
CAS Number: 776-34-1
MDL Number: MFCD00004026
Molecular Formula: C10H8N2O2
Molecular Weight: 188.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 195
Formula:
C₁₀H₈N₂O₂ Melting Pt: 195 °C |
MDL Number:
MFCD00004026 CAS Number: 776-34-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...