2-Nitro-9-fluorenone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Nitro-9H-fluoren-9-one

N0202-10G N0202-1G
TCN0202-010GEA 142.2 CAD
TCN0202-010G TCN0202-001G
2-Nitro-9-fluorenone ≥98.0% (by HPLC)
2-Nitro-9-fluorenone

CAS Number: 3096-52-4
MDL Number: MFCD00001152
Molecular Formula: C13H7NO3
Molecular Weight: 225.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 222

Formula: C₁₃H₇NO₃
MW: 225.2 g/mol
Boiling Pt: 419 °C (1013 hPa)
Melting Pt: 222 °C
Density: 1.44 g/cm³ (20 °C)
MDL Number: MFCD00001152
CAS Number: 3096-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f5b326e3deaac65ebcd08254a47d3cce8c3a90d&vid=7f5b326e3deaac65ebcd08254a47d3cce8c3a90d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR