5-Nitrofluorescein (isomer I) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Nitro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, 3',6'-Dihydroxy-5-nitro-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

N0199-5G N0199-1G
TCN0199-005GEA 1099.68 CAD
TCN0199-005G TCN0199-001G
5-Nitrofluorescein (isomer I) ≥98.0% (by titrimetric analysis)
5-Nitrofluorescein (isomer I)

CAS Number: 3326-35-0
MDL Number: MFCD00135108
Molecular Formula: C20H11NO7
Molecular Weight: 377.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 5-Nitro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one

Formula: C₂₀H₁₁NO₇
MW: 377.31 g/mol
MDL Number: MFCD00135108
CAS Number: 3326-35-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0d5f0346a4c3b043124651098d07723cc6ee0d7&vid=e0d5f0346a4c3b043124651098d07723cc6ee0d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR