4'-Nitro-4-dimethylaminoazobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4-Dimethylamino-4'-nitroazobenzene

N0192-25G N0192-5G
TCN0192-025GEA 663.6 CAD
TCN0192-025G TCN0192-5G
4'-Nitro-4-dimethylaminoazobenzene ≥98.0%
4'-Nitro-4-dimethylaminoazobenzene

CAS Number: 2491-74-9
MDL Number: MFCD00059834
Molecular Formula: C14H14N4O2
Molecular Weight: 270.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 229


Other Product Names / Synonyms: 4-Dimethylamino-4'-nitroazobenzene

Formula: C₁₄H₁₄N₄O₂
Melting Pt: 229 °C
MDL Number: MFCD00059834
CAS Number: 2491-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=659107e0f61ba358c2bd542891999bf7928316ee&vid=659107e0f61ba358c2bd542891999bf7928316ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR