5-Nitrobenzimidazole nitrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0151-25G N0151-250G
TCN0151-25GEA 63.2 CAD
TCN0151-25G TCN0151-250G
5-Nitrobenzimidazole nitrate ≥98.0% (by titrimetric analysis)
5-Nitrobenzimidazole nitrate

CAS Number: 27896-84-0
MDL Number: MFCD00013157
Molecular Formula: C7H5N3O2
Molecular Weight: 226.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 222

Formula: C₇H₅N₃O₂
Melting Pt: 222 °C
MDL Number: MFCD00013157
CAS Number: 27896-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d599736c658ae89681eb14369300b0d3625e9af9&vid=d599736c658ae89681eb14369300b0d3625e9af9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR