1-Nitro-9,10-anthraquinone ≥98.0%

Supplier: TCI America

N0127-25G N0127-500G
TCN0127-025GEA 164.32 CAD
TCN0127-025G TCN0127-500G
1-Nitro-9,10-anthraquinone ≥98.0%
1-Nitro-9,10-anthraquinone

CAS Number: 82-34-8
MDL Number: MFCD00019140
Molecular Formula: C14H7NO4
Molecular Weight: 253.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 232

Formula: C₁₄H₇NO₄
Melting Pt: 232 °C
MDL Number: MFCD00019140
CAS Number: 82-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c4add5681d5b179854e8659b79bee36104ce142&vid=1c4add5681d5b179854e8659b79bee36104ce142&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR