Triethanolamine phosphate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
N0103-25G
N0103-500G
TCN0103-025GEA
87.53
CAD
TCN0103-025G
TCN0103-500G
Triethanolamine phosphate ≥98.0% (by titrimetric analysis)
Triethanolamine phosphate
CAS Number: 10017-56-8
MDL Number: MFCD00054303
Molecular Formula: C6H15NO3
Molecular Weight: 247.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 106
Other Product Names / Synonyms: 2,2',2''-Nitrilotriethanol Phosphate
Formula:
C₆H₁₅NO₃ Melting Pt: 106 °C |
MDL Number:
MFCD00054303 CAS Number: 10017-56-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...