Nitrilotriacetic acid trisodium salt monohydrate ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
Nitrilotriacetic acid, trisodium salt, monohydrate
N0101-500G
N0101-25G
TCN0101-500GEA
74.28
CAD
TCN0101-500G
TCN0101-025G
Nitrilotriacetic acid trisodium salt monohydrate ≥98.0% for synthesis
Nitrilotriacetic acid trisodium salt monohydrate
CAS Number: 18662-53-8
MDL Number: MFCD00150789
Molecular Formula: C6H9NO6
Molecular Weight: 257.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Triglycollamic Acid Trisodium Salt, Nitrilotriacetic Acid Trisodium Salt, NTA 3Na
Formula:
C₆H₆NNa₃O₆·1H₂O MW: 275.1 g/mol |
MDL Number:
MFCD00150789 CAS Number: 18662-53-8 EINECS: 225-768-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...