Nipecotamide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
N0097-5G
N0097-25G
TCN0097-005GEA
82.16
CAD
TCN0097-005G
TCN0097-025G
Nipecotamide ≥98.0% (by titrimetric analysis)
Nipecotamide
CAS Number: 4138-26-5
MDL Number: MFCD00005993
Molecular Formula: C6H12N2O
Molecular Weight: 128.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 111
Other Product Names / Synonyms: 3-Piperidinecarboxamide
Formula:
C₆H₁₂N₂O Melting Pt: 111 °C |
MDL Number:
MFCD00005993 CAS Number: 4138-26-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...