(±)-Naringenin ≥93.0% (by HPLC, titration analysis)

Supplier: TCI America

N0072-25G N0072-5G
TCN0072-025GEA 314.42 CAD
TCN0072-025G TCN0072-5G
(±)-Naringenin ≥93.0% (by HPLC, titration analysis)
(±)-Naringenin

CAS Number: 67604-48-2
MDL Number: MFCD00006844
Molecular Formula: C15H12O5
Molecular Weight: 272.26
Purity/Analysis Method: >93.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 4',5,7-Trihydroxyflavanone

Formula: C₁₅H₁₂O₅
MDL Number: MFCD00006844
CAS Number: 67604-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b52583d821a634898b42209c2b9d5f114c92cacf&vid=b52583d821a634898b42209c2b9d5f114c92cacf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR