1-Acetamidonaphthalene ≥99.0%

Supplier: TCI America

N0050-250G N0050-25G
TCN0050-250GEA 325.48 CAD
TCN0050-250G TCN0050-025G
1-Acetamidonaphthalene ≥99.0%
1-Acetamidonaphthalene

CAS Number: 575-36-0
MDL Number: MFCD00059128
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: 1-Acetonaphthalide, N-Acetyl-1-naphthylamine, N-1-Naphthylacetamide

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00059128
CAS Number: 575-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b622bda7386eb363b35b54af879e44edc265e43&vid=2b622bda7386eb363b35b54af879e44edc265e43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR