2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione ≥98.0%
Supplier: TCI America
A1853-1G
A1853-5G
TCA1853-1GEA
71.1
CAD
TCA1853-1G
TCA1853-5G
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione ≥98.0%
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione
CAS Number: 1755-15-3
Molecular Formula: C10H14O3
Molecular Weight: 182.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 36
Other Product Names / Synonyms: 2-Acetyldimedone
Formula:
C₁₀H₁₄O₃ Melting Pt: 36 °C |
MDL Number:
MFCD00467663 CAS Number: 1755-15-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...