Sodium-2-naphthol-6-sulfonate hydrate ≥97.0%

Supplier: TCI America

Synonyms: Sodium 6-hydroxynaphthalene-2-sulphonate, 6-Hydroxy-2-naphthalenesulphonic acid Sodium Salt, 2-Naphthol-6-sulphonic acid Sodium Salt, 6-Hydroxy-2-naphthalenesulfonic acid Sodium Salt, Sodium 6-hydroxynaphthalene-2-sulfonate, Sodium 6-Hydroxy-2-naphthalenesulfonate, Sodium 6-Hydroxy-2-naphthalenesulphonate, 2-Naphthol-6-sulfonic acid Sodium Salt, Schaeffer's acid Sodium Salt

N0035-25G N0035-500G
TCN0035-025GEA 55.3 CAD
TCN0035-025G TCN0035-500G
Sodium-2-naphthol-6-sulfonate hydrate ≥97.0%
Sodium-2-naphthol-6-sulfonate hydrate

CAS Number: 135-76-2
MDL Number: MFCD00070488
Molecular Formula: C10H8O4S
Molecular Weight: 246.21
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Hydroxy-2-naphthalenesulfonic Acid Sodium Salt, Schaeffer's Acid Sodium Salt, Sodium 6-Hydroxy-2-naphthalenesulfonate, 2-Naphthol-6-sulfonic Acid Sodium Salt

Formula: C₁₀H₈O₄S
MW: 246.21 g/mol
MDL Number: MFCD00070488
CAS Number: 135-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a6f877ced493c99e1c5b8ed89412a837c089ddc&vid=1a6f877ced493c99e1c5b8ed89412a837c089ddc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR