Sodium-1-naphthol-8-sulfonate ≥96.0% (by HPLC)

Supplier: TCI America

N0034-25G N0034-500G
TCN0034-025GEA 28.44 CAD
TCN0034-025G TCN0034-500G
Sodium-1-naphthol-8-sulfonate ≥96.0% (by HPLC)
Sodium-1-naphthol-8-sulfonate

CAS Number: 20215-36-5
MDL Number: MFCD00021524
Molecular Formula: C10H8O4S
Molecular Weight: 246.21
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 8-Hydroxy-1-naphthalenesulfonic Acid Sodium Salt, Sodium 8-Hydroxy-1-naphthalenesulfonate, 1-Naphthol-8-sulfonic Acid Sodium Salt

Formula: C₁₀H₈O₄S
MDL Number: MFCD00021524
CAS Number: 20215-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98d58dac28ec8a1409580f780bb5bd7e968d539b&vid=98d58dac28ec8a1409580f780bb5bd7e968d539b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR