Sodium-1-naphthol-4-sulfonate ≥85.0%
Supplier: TCI America
Synonyms:
4-Hydroxy-1-naphthalenesulphonic acid Sodium Salt, Sodium 4-Hydroxy-1-naphthalenesulphonate, 4-Hydroxy-1-naphthalenesulfonic acid Sodium Salt, Neville Winther acid, Sodium 4-Hydroxy-1-naphthalenesulfonate, 1-Naphthol-4-sulfonic acid Sodium Salt, NW acid, 1-Naphthol-4-sulphonic acid Sodium Salt
CAS Number: 6099-57-6
MDL Number: MFCD00021518
Molecular Formula: C10H8O4S
Molecular Weight: 246.21
Purity/Analysis Method: >85.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 4-Hydroxy-1-naphthalenesulfonic Acid Sodium Salt, 1-Naphthol-4-sulfonic Acid Sodium Salt, Neville Winther Acid, NW Acid, Sodium 4-Hydroxy-1-naphthalenesulfonate
Formula:
C₁₀H₈O₄S MW: 246.21 g/mol |
MDL Number:
MFCD00021518 CAS Number: 6099-57-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...