Trisodium naphthalene-1,3,6-trisulfonate hydrate 65.0% (by HPLC)

Supplier: TCI America

Synonyms: Naphthalene-1,3,6-trisulphonic acid Trisodium Salt, Naphthalene-1,3,6-trisulfonic acid Trisodium Salt

N0021-25G N0021-500G N0021-100G
TCN0021-025GEA 56.25 CAD
TCN0021-025G TCN0021-500G TCN0021-100G
Trisodium naphthalene-1,3,6-trisulfonate hydrate 65.0% (by HPLC)
Trisodium naphthalene-1,3,6-trisulfonate hydrate

CAS Number: 5182-30-9
MDL Number: MFCD00003989
Molecular Formula: C10H8O9S3
Molecular Weight: 434.29
Purity/Analysis Method: >65.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Naphthalene-1,3,6-trisulfonic Acid Trisodium Salt

Formula: C₁₀H₈O₉S₃·xH₂O
MW: 434.29 g/mol
MDL Number: MFCD00003989
CAS Number: 5182-30-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a76e2150aa9c7c0bb00ac56b0956477b37d5a8c&vid=2a76e2150aa9c7c0bb00ac56b0956477b37d5a8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 65.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR