Disodium-2,6-naphthalenedisulfonate ≥90.0%

Supplier: TCI America

Synonyms: 2,6-Naphthalenedisulfonic acid disodium salt, Sodium naphthalene-2,6-disulfonate, Sodium naphthalene-2,6-disulphonate, Ebert-Merz beta-acid disodium salt, 2,6-Naphthalenedisulphonic acid disodium salt

N0013-500G N0013-25G
TCN0013-500GEA 461.36 CAD
TCN0013-500G TCN0013-025G
Disodium-2,6-naphthalenedisulfonate ≥90.0%
Disodium-2,6-naphthalenedisulfonate

CAS Number: 1655-45-4
MDL Number: MFCD00004092
Molecular Formula: C10H8O6S2
Molecular Weight: 332.25
Purity/Analysis Method: >90.0% (T)
Form: Crystal


Other Product Names / Synonyms: Ebert-Merz beta-Acid Disodium Salt, 2,6-Naphthalenedisulfonic Acid Disodium Salt

Formula: C₁₀H₈O₆S₂
MDL Number: MFCD00004092
CAS Number: 1655-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6201bdfa44843daf0e1c219ef832674958bba773&vid=6201bdfa44843daf0e1c219ef832674958bba773&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR