1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M2098-25G M2098-5G
TCM2098-25GEA 461.36 CAD
TCM2098-25G TCM2098-5G
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulfonyl)imide

CAS Number: 223437-05-6
MDL Number: MFCD08458917
Molecular Formula: C10H18F6N2O4S2
Molecular Weight: 408.37
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Melting point (°C): 12
Specific Gravity (20/20): 1.44

Formula: C₁₀H₁₈F₆N₂O₄S₂
Melting Pt: 12 °C
Density: 1.44 (20 °C)
MDL Number: MFCD08458917
CAS Number: 223437-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae71a25f161cb8c9e9159c3fb54d3bfcfef64745&vid=ae71a25f161cb8c9e9159c3fb54d3bfcfef64745&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR