4-Methoxy-1-naphthalenemethanol ≥96.0%

Supplier: TCI America

Synonyms: 1-(Hydroxymethyl)-4-methoxynaphthalene, (4-Methoxynaphthalen-1-yl)methanol

M2086-1G M2086-5G
TCM2086-1GEA 167.48 CAD
TCM2086-1G TCM2086-5G
4-Methoxy-1-naphthalenemethanol ≥96.0%
4-Methoxy-1-naphthalenemethanol

CAS Number: 16820-54-5
MDL Number: MFCD04089264
Molecular Formula: C12H12O2
Molecular Weight: 188.23
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 76


Other Product Names / Synonyms: 1-(Hydroxymethyl)-4-methoxynaphthalene

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Melting Pt: 76 °C
MDL Number: MFCD04089264
CAS Number: 16820-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18edd0ecfddadb5079e4c0f504d9547c3a20465a&vid=18edd0ecfddadb5079e4c0f504d9547c3a20465a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR