Methyl-4-amino-3-bromobenzoate ≥98.0%

Supplier: TCI America

M2084-1G M2084-5G
TCM2084-1GEA 41.08 CAD
TCM2084-1G TCM2084-5G
Methyl-4-amino-3-bromobenzoate ≥98.0%
Methyl 4-amino-3-bromobenzoate

CAS Number: 106896-49-5
MDL Number: MFCD01861385
Molecular Formula: C8H8BrNO2
Molecular Weight: 230.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 107


Other Product Names / Synonyms: 4-Amino-3-bromobenzoic Acid Methyl Ester

Formula: C₈H₈BrNO₂
Melting Pt: 107 °C
MDL Number: MFCD01861385
CAS Number: 106896-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32f283dbbf247306eb912f73d0f769a53ed825a7&vid=32f283dbbf247306eb912f73d0f769a53ed825a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR