Methyl-5-formylsalicylate 98,0

Supplier: TCI America

Synonyms: Methyl 5-Formyl-2-hydroxybenzoate

M2082-25G M2082-5G
TCM2082-25GEA 219.62 CAD
TCM2082-25G TCM2082-5G
Methyl-5-formylsalicylate 98,0
Methyl 5-formylsalicylate

CAS Number: 41489-76-3
MDL Number: MFCD03840784
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 81


Other Product Names / Synonyms: 5-Formylsalicylic Acid Methyl Ester, Methyl 5-Formyl-2-hydroxybenzoate, 5-Formyl-2-hydroxybenzoic Acid Methyl Ester

Formula: C₉H₈O₄
MW: 180.1574 g/mol
Melting Pt: 81 °C
MDL Number: MFCD03840784
CAS Number: 41489-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5a89e123d8b4fe2581df83e2d63e63e01b0177b&vid=c5a89e123d8b4fe2581df83e2d63e63e01b0177b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR