ɑ-Methyl-L-proline ≥96.0% (by titrimetric analysis)

Supplier: TCI America

M2077-1G M2077-5G
TCM2077-1GEA 457.88 CAD
TCM2077-1G TCM2077-5G
ɑ-Methyl-L-proline ≥96.0% (by titrimetric analysis)
alpha-Methyl-L-proline

CAS Number: 42856-71-3
MDL Number: MFCD01318647
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 310
Specific rotation [a]20/D: -73 deg (C=1, MeOH)

Formula: C₆H₁₁NO₂
Melting Pt: 310 °C
MDL Number: MFCD01318647
CAS Number: 42856-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a45517372525fcd29d98d778fbcb47ea92504cf&vid=5a45517372525fcd29d98d778fbcb47ea92504cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR