4-Methoxy-4'-methyldiphenylamine ≥98.0%

Supplier: TCI America

Synonyms: 4-Methoxy-N-(p-tolyl)aniline, N-(4-Methoxyphenyl)-p-tolylamine

M2070-25G M2070-5G
TCM2070-25GEA 467.68 CAD
TCM2070-25G TCM2070-5G
4-Methoxy-4'-methyldiphenylamine ≥98.0%
4-Methoxy-4'-methyldiphenylamine

CAS Number: 39253-43-5
MDL Number: MFCD00092922
Molecular Formula: C14H15NO
Molecular Weight: 213.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 84


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: N-(4-Methoxyphenyl)-p-tolylamine

Formula: C₁₄H₁₅NO
MW: 213.28 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00092922
CAS Number: 39253-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=856d3f923ca28c61fa201157ea06de6d01efd285&vid=856d3f923ca28c61fa201157ea06de6d01efd285&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR