4-Methoxydiphenylamine ≥98.0%
Supplier: TCI America
M2069-25G
M2069-5G
TCM2069-25GEA
1082.3
CAD
TCM2069-25G
TCM2069-5G
4-Methoxydiphenylamine ≥98.0%
4-Methoxydiphenylamine
CAS Number: 1208-86-2
MDL Number: MFCD00228649
Molecular Formula: C13H13NO
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 195
Melting point (°C): 106
Formula:
C₁₃H₁₃NO Boiling Pt: 195 °C Melting Pt: 106 °C |
MDL Number:
MFCD00228649 CAS Number: 1208-86-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...