Methyl-2-(bromomethyl)acrylate ≥97.0%

Supplier: TCI America

M2067-5G M2067-1G
TCM2067-5GEA 162.74 CAD
TCM2067-5G TCM2067-1G
Methyl-2-(bromomethyl)acrylate ≥97.0%
Methyl-2-(bromomethyl)acrylate

CAS Number: 4224-69-5
MDL Number: MFCD00011697
Molecular Formula: C5H7BrO2
Molecular Weight: 179.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 42
Flash Point (°C): 78
Specific Gravity (20/20): 1.50
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(Bromomethyl)acrylic Acid Methyl Ester

Formula: C₅H₇BrO₂
Boiling Pt: 42 °C
Density: 1.50 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00011697
CAS Number: 4224-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d16fba3fd1debab37bb6cdeccfd80de32df6890d&vid=d16fba3fd1debab37bb6cdeccfd80de32df6890d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR