1-Methyl-3-octylimidazolium chloride ≥98.0%

Supplier: TCI America

Synonyms: 1-Methyl-3-n-octylimidazolium chloride

M2062-5G M2062-25G
TCM2062-5GEA 80.58 CAD
TCM2062-5G TCM2062-25G
1-Methyl-3-octylimidazolium chloride ≥98.0%
1-Methyl-3-octylimidazolium chloride

CAS Number: 64697-40-1
MDL Number: MFCD03095432
Molecular Formula: C12H23ClN2
Molecular Weight: 230.78
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Melting point (°C): 12

Formula: C₁₂H₂₃ClN₂
Melting Pt: 12 °C
MDL Number: MFCD03095432
CAS Number: 64697-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29b33594c8458e1cb4a99a2f1683d5e580d88e38&vid=29b33594c8458e1cb4a99a2f1683d5e580d88e38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR