Methyl-2,3:4,6-di-O-benzylidene-ɑ-D-mannopyranoside ≥98.0%

Supplier: TCI America

M2061-25G M2061-5G
TCM2061-25GEA 824.76 CAD
TCM2061-25G TCM2061-5G
Methyl-2,3:4,6-di-O-benzylidene-ɑ-D-mannopyranoside ≥98.0%
Methyl-2,3:4,6-di-O-benzylidene-alpha-D-mannopyranoside

CAS Number: 4148-71-4
MDL Number: MFCD00192286
Molecular Formula: C21H22O6
Molecular Weight: 370.40
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 185

Formula: C₂₁H₂₂O₆
MW: 370.40 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00192286
CAS Number: 4148-71-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fcc33cbac298ea1813e560526aedc7418b6ad1e&vid=6fcc33cbac298ea1813e560526aedc7418b6ad1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR