3-Methoxybutyl thioglycolate ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M2060-25G
M2060-500G
TCM2060-25GEA
47.4
CAD
TCM2060-25G
TCM2060-500G
3-Methoxybutyl thioglycolate ≥97.0% (by GC, titration analysis)
3-Methoxybutyl thioglycolate
CAS Number: 27431-39-6
MDL Number: MFCD04038084
Molecular Formula: C7H14O3S
Molecular Weight: 178.25
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: Thioglycolic Acid 3-Methoxybutyl Ester, 3-Methoxybutyl Mercaptoacetate, Mercaptoacetic Acid 3-Methoxybutyl Ester
Formula:
C₇H₁₄O₃S Density: 1.08 (20 °C) |
MDL Number:
MFCD04038084 CAS Number: 27431-39-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...