6-Methoxy-8-nitroquinoline ≥98.0%

Supplier: TCI America

M2057-25G M2057-5G
TCM2057-25GEA 588.39 CAD
TCM2057-25G TCM2057-5G
6-Methoxy-8-nitroquinoline ≥98.0%
6-Methoxy-8-nitroquinoline

CAS Number: 85-81-4
MDL Number: MFCD00006802
Molecular Formula: C10H8N2O3
Molecular Weight: 204.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 161

Formula: C₁₀H₈N₂O₃
Melting Pt: 161 °C
MDL Number: MFCD00006802
CAS Number: 85-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a211edddf22210d4d5ca81f10c13fb9d5ec81b5d&vid=a211edddf22210d4d5ca81f10c13fb9d5ec81b5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR