2-Anthraceneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2-Anthraceneboronic acid, Anthracen-2-ylboronic acid, 2-Anthraceneboronic Acid

A1843-1G A1843-5G
TCA1843-1GEA 243.32 CAD
TCA1843-1G TCA1843-5G
2-Anthraceneboronic acid (contains varying amounts of Anhydride)
2-Anthraceneboronic acid

CAS Number: 141981-64-8
MDL Number: MFCD07784001
Molecular Formula: C14H11BO2
Molecular Weight: 222.05
Form: Crystal
Color: Very Pale Yellow

Formula: C₁₄H₁₁BO₂
MW: 222.05 g/mol
MDL Number: MFCD07784001
CAS Number: 141981-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c755f9bd10b92e5ad7d7a164e07be64097528c09&vid=c755f9bd10b92e5ad7d7a164e07be64097528c09&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR