(4-Chlorophenyl)acetic acid methyl ester ≥98.0%

Supplier: TCI America

M2039-5G M2039-25G
TCM2039-5GEA 44.24 CAD
TCM2039-5G TCM2039-25G
(4-Chlorophenyl)acetic acid methyl ester ≥98.0%
(4-Chlorophenyl)acetic acid methyl ester

CAS Number: 52449-43-1
MDL Number: MFCD00032743
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: (4-Chlorophenyl)acetic Acid Methyl Ester

Formula: (C₆H₄Cl)CH₂COOCH₃
Boiling Pt: 115 °C
Density: 1.20 (20 °C)
MDL Number: MFCD00032743
CAS Number: 52449-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b940b3d82baa48b17cc6dc62e3ecdb02fbc299&vid=21b940b3d82baa48b17cc6dc62e3ecdb02fbc299&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR