4-(4-Methoxyphenyl)butyric acid ≥95.0%
Supplier: TCI America
M2037-5G
M2037-25G
TCM2037-5GEA
121.66
CAD
TCM2037-5G
TCM2037-25G
4-(4-Methoxyphenyl)butyric acid ≥95.0%
4-(4-Methoxyphenyl)butyric acid
CAS Number: 4521-28-2
MDL Number: MFCD00004404
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 196
Melting point (°C): 60
Formula:
C₁₁H₁₄O₃ Boiling Pt: 196 °C Melting Pt: 60 °C |
MDL Number:
MFCD00004404 CAS Number: 4521-28-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...