5-Nitro-2-picoline ≥96.0%

Supplier: TCI America

Synonyms: 2-Methyl-5-nitropyridine

M2034-1G M2034-5G
TCM2034-1GEA 52.14 CAD
TCM2034-1G TCM2034-5G
5-Nitro-2-picoline ≥96.0%
5-Nitro-2-picoline

CAS Number: 21203-68-9
MDL Number: MFCD04114179
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 109

Formula: C₆H₆N₂O₂
Melting Pt: 109 °C
MDL Number: MFCD04114179
CAS Number: 21203-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa5a552db9af174a7a17bda77807bf289d74b432&vid=aa5a552db9af174a7a17bda77807bf289d74b432&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR