(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride ≥95.0%

Supplier: TCI America

Synonyms: H-Asp(OMe)-OH·HCl, L-Aspartic acid 4-methyl ester hydrochloride, L-Aspartatic Acid 4-Methyl Ester Hydrochloride, L-Aspartic acid β-methyl ester hydrochloride, H-Asp(OMe)-OH.HCl, H-Asp(OMe)-OH β-Methyl L-aspartate hydrochloride

M2024-5G M2024-25G
TCM2024-5GEA 64.78 CAD
TCM2024-5G TCM2024-25G
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride ≥95.0%
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride

CAS Number: 16856-13-6
MDL Number: MFCD00038972
Molecular Formula: C5H9NO4
Molecular Weight: 183.59
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 193
Specific rotation [a]20/D: 11 deg (C=1, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free), Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Aspartatic Acid 4-Methyl Ester Hydrochloride, H-Asp(OMe)-OH.HCl

Formula: C₅H₉NO₄·HCl
MW: 183.6 g/mol
Melting Pt: 193 °C
MDL Number: MFCD00038972
CAS Number: 16856-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1c410b43daa9f8fb9b8df8e797e9f90affd6d3d&vid=e1c410b43daa9f8fb9b8df8e797e9f90affd6d3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR