2-Methoxy-1-naphthalenemethanol ≥98.0%
Supplier: TCI America
M2018-25G
M2018-5G
TCM2018-25GEA
508.76
CAD
TCM2018-25G
TCM2018-5G
2-Methoxy-1-naphthalenemethanol ≥98.0%
2-Methoxy-1-naphthalenemethanol
CAS Number: 40696-22-8
MDL Number: MFCD00274224
Molecular Formula: C12H12O2
Molecular Weight: 188.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103
Other Product Names / Synonyms: 1-(Hydroxymethyl)-2-methoxynaphthalene
Formula:
C₁₂H₁₂O₂ Melting Pt: 103 °C |
MDL Number:
MFCD00274224 CAS Number: 40696-22-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...