4-Methoxybenzyl 2,2,2-trichloroacetimidate ≥96.0%
Supplier: TCI America
Synonyms:
2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
CAS Number: 89238-99-3
MDL Number: MFCD00134547
Molecular Formula: C10H10Cl3NO2
Molecular Weight: 282.55
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 110
Specific Gravity (20/20): 1.36
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
Formula:
C₁₀H₁₀Cl₃NO₂ MW: 282.55 g/mol Boiling Pt: 137 °C Density: 1.36 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00134547 CAS Number: 89238-99-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...